Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCR3
Entrez Gene ID
259197
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
1C7, CD337, LY117, MALS, NKp30
Chromosome Location
Chr.6: 31588883 - 31593024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145467.1 NP_001138939.1 3 - 4 761 55
GenBank mRNA AB055881.1 - 3 - 4 497 55
AF031138.1 - 3 - 4 761 55
AJ223153.1 - 3 - 4 560 55

View Full Product Details