The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000428.2 | NP_000419.1 | 30 - 31 | 4843 | 60 |
XM_011536765.1 | XP_011535067.1 | 28 - 29 | 4370 | 60 | |
GenBank mRNA | AB209865.1 | - | 30 - 31 | 4093 | 60 |
BC078659.1 | - | 31 - 32 | 4749 | 60 | |
S82451.1 | - | 30 - 31 | 4843 | 60 | |
Z37976.1 | - | 30 - 31 | 4844 | 60 |