Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYBB
Entrez Gene ID
1536
Gene Name
cytochrome b-245 beta chain
Gene Aliases
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX
Chromosome Location
Chr.X: 37780017 - 37813461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000397.3 NP_000388.2 3 - 4 315 78
GenBank mRNA AK289753.1 - 3 - 4 300 78
AK301491.1 - 2 - 3 177 78
AK304033.1 - 3 - 4 275 78
AK312929.1 - 3 - 4 275 78
BC032720.1 - 3 - 4 301 78
DA995835.1 - 3 - 4 315 78
KU178009.1 - 3 - 4 254 78
KU178010.1 - 3 - 4 254 78
X04011.1 - 3 - 4 269 78

View Full Product Details