The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033272.3 | NP_150375.2 | 13 - 14 | 3191 | 85 |
XM_011512109.2 | XP_011510411.1 | 14 - 15 | 3105 | 85 | |
XM_017005218.1 | XP_016860707.1 | 14 - 15 | 3096 | 85 | |
XM_017005219.1 | XP_016860708.1 | 13 - 14 | 3072 | 85 | |
XM_017005220.1 | XP_016860709.1 | 12 - 13 | 3060 | 85 | |
GenBank mRNA | AF032897.1 | - | 13 - 14 | 3062 | 85 |