The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033272.3 | NP_150375.2 | 5 - 6 | 1138 | 65 |
XM_017005219.1 | XP_016860708.1 | 5 - 6 | 1028 | 65 | |
GenBank mRNA | AF032897.1 | - | 5 - 6 | 1009 | 65 |