The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005768.5 | NP_005759.4 | 10 - 11 | 1303 | 109 |
GenBank mRNA | AK058063.1 | - | 4 - 5 | 585 | 109 |
AK096775.1 | - | 10 - 11 | 1491 | 109 | |
AK296145.1 | - | 9 - 10 | 1013 | 109 | |
AK315538.1 | - | 8 - 9 | 875 | 109 | |
BC000664.2 | - | 9 - 10 | 1038 | 109 | |
BC065194.1 | - | 10 - 11 | 1234 | 109 | |
BT007000.1 | - | 8 - 9 | 871 | 109 | |
BX648009.1 | - | 10 - 11 | 1274 | 109 | |
U72515.1 | - | 9 - 10 | 988 | 109 |