The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005768.5 | NP_005759.4 | 5 - 6 | 611 | 79 |
GenBank mRNA | AK096775.1 | - | 5 - 6 | 800 | 79 |
AK296145.1 | - | 4 - 5 | 321 | 79 | |
AK311727.1 | - | 3 - 4 | 333 | 79 | |
AK315538.1 | - | 3 - 4 | 183 | 79 | |
BC000664.2 | - | 4 - 5 | 346 | 79 | |
BC065194.1 | - | 5 - 6 | 542 | 79 | |
BT007000.1 | - | 3 - 4 | 179 | 79 | |
BX647983.1 | - | 5 - 6 | 551 | 79 | |
BX648009.1 | - | 5 - 6 | 582 | 79 | |
U72515.1 | - | 4 - 5 | 296 | 79 |