The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178229.4 | NP_839943.2 | 18 - 19 | 2210 | 115 |
XM_011509198.2 | XP_011507500.1 | 18 - 19 | 2199 | 115 | |
XM_011509200.2 | XP_011507502.1 | 7 - 8 | 1379 | 115 | |
XM_011509201.2 | XP_011507503.1 | 6 - 7 | 768 | 115 | |
XM_017000317.1 | XP_016855806.1 | 18 - 19 | 2196 | 115 | |
XM_017000318.1 | XP_016855807.1 | 16 - 17 | 1953 | 115 | |
GenBank mRNA | AB105103.1 | - | 18 - 19 | 2146 | 115 |
AK122653.1 | - | 4 - 5 | 418 | 115 | |
AY253300.1 | - | 18 - 19 | 2135 | 115 | |
BC131536.1 | - | 18 - 19 | 2168 | 115 |