The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178229.4 | NP_839943.2 | 22 - 23 | 2607 | 71 |
XM_011509198.2 | XP_011507500.1 | 22 - 23 | 2596 | 71 | |
XM_011509200.2 | XP_011507502.1 | 11 - 12 | 1776 | 71 | |
XM_011509201.2 | XP_011507503.1 | 10 - 11 | 1165 | 71 | |
XM_017000317.1 | XP_016855806.1 | 22 - 23 | 2593 | 71 | |
XM_017000318.1 | XP_016855807.1 | 20 - 21 | 2350 | 71 | |
GenBank mRNA | AB105103.1 | - | 22 - 23 | 2543 | 71 |
AK122653.1 | - | 8 - 9 | 815 | 71 | |
AY253300.1 | - | 22 - 23 | 2532 | 71 | |
BC131536.1 | - | 22 - 23 | 2565 | 71 |