The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 12 - 13 | 2133 | 139 |
GenBank mRNA | AF395908.1 | - | 12 - 13 | 1793 | 139 |
AF395909.1 | - | 12 - 13 | 1793 | 139 | |
AF411577.1 | - | 12 - 13 | 1793 | 139 | |
AF411578.1 | - | 12 - 13 | 1793 | 139 | |
AY040220.1 | - | 12 - 13 | 2133 | 139 | |
AY196326.1 | - | 11 - 12 | 1839 | 139 |