The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 18 - 19 | 2828 | 71 |
GenBank mRNA | AF395908.1 | - | 18 - 19 | 2479 | 71 |
AF395909.1 | - | 18 - 19 | 2479 | 71 | |
AF411577.1 | - | 18 - 19 | 2479 | 71 | |
AF411578.1 | - | 18 - 19 | 2479 | 71 | |
AY040220.1 | - | 18 - 19 | 2828 | 71 | |
AY196326.1 | - | 16 - 17 | 2462 | 71 | |
BC036378.1 | - | 19 - 20 | 2904 | 71 |