The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 20 - 21 | 3250 | 91 |
GenBank mRNA | AF395908.1 | - | 20 - 21 | 2901 | 91 |
AF411577.1 | - | 20 - 21 | 2901 | 91 | |
AF411578.1 | - | 20 - 21 | 2901 | 91 | |
AY040220.1 | - | 20 - 21 | 3250 | 91 | |
AY196326.1 | - | 18 - 19 | 2884 | 91 | |
BC036378.1 | - | 21 - 22 | 3326 | 91 |