The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033226.2 | NP_150229.2 | 3 - 4 | 772 | 76 |
GenBank mRNA | AY040220.1 | - | 3 - 4 | 772 | 76 |