The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002159.3 | NP_002150.1 | 2 - 3 | 146 | 128 |
GenBank mRNA | BC017835.1 | - | 2 - 3 | 119 | 128 |
BP335073.2 | - | 2 - 3 | 123 | 128 | |
CB984538.1 | - | 2 - 3 | 150 | 128 | |
CR627329.1 | - | 1 - 2 | 248 | 206 | |
M26664.1 | - | 1 - 2 | 61 | 128 |