The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011722.2 | NP_001011722.2 | 24 - 25 | 2936 | 131 |
NM_001319837.1 | NP_001306766.1 | 23 - 24 | 2921 | 131 | |
NM_001319838.1 | NP_001306767.1 | 15 - 16 | 2160 | 131 | |
NM_001328124.1 | NP_001315053.1 | 18 - 19 | 2385 | 131 | |
NM_018125.3 | NP_060595.3 | 26 - 27 | 3169 | 131 | |
NR_137287.1 | 27 - 28 | 3916 | 131 | ||
NR_137288.1 | 26 - 27 | 3796 | 131 | ||
XM_005245923.2 | XP_005245980.2 | 25 - 26 | 3028 | 131 | |
XM_005245925.2 | XP_005245982.2 | 25 - 26 | 3016 | 131 | |
XM_005245927.2 | XP_005245984.2 | 18 - 19 | 2385 | 131 | |
XM_005245929.2 | XP_005245986.2 | 17 - 18 | 2370 | 131 | |
XM_006710728.1 | XP_006710791.1 | 25 - 26 | 3210 | 131 | |
XM_006710729.1 | XP_006710792.1 | 25 - 26 | 3025 | 131 | |
XM_006710731.1 | XP_006710794.1 | 25 - 26 | 3207 | 131 | |
XM_011541691.2 | XP_011539993.1 | 24 - 25 | 3025 | 131 | |
XM_011541692.2 | XP_011539994.1 | 23 - 24 | 3019 | 131 | |
XM_011541693.2 | XP_011539995.1 | 23 - 24 | 3010 | 131 | |
XM_017001617.1 | XP_016857106.1 | 24 - 25 | 3001 | 131 | |
XM_017001618.1 | XP_016857107.1 | 19 - 20 | 2303 | 131 | |
XM_017001619.1 | XP_016857108.1 | 24 - 25 | 3034 | 131 | |
XM_017001620.1 | XP_016857109.1 | 23 - 24 | 3019 | 131 | |
XM_017001621.1 | XP_016857110.1 | 25 - 26 | 3152 | 131 | |
XM_017001622.1 | XP_016857111.1 | 24 - 25 | 3137 | 131 | |
GenBank mRNA | AB046846.1 | - | 26 - 27 | 3169 | 131 |
AK092020.1 | - | 15 - 16 | 2157 | 131 | |
AK126877.1 | - | 17 - 18 | 3275 | 131 | |
AK300264.1 | - | 17 - 18 | 2333 | 131 | |
AK311076.1 | - | 25 - 26 | 2979 | 131 | |
BC065561.1 | - | 12 - 13 | 1671 | 131 | |
BC080596.1 | - | 15 - 16 | 1832 | 131 | |
BC117171.1 | - | 24 - 25 | 2926 | 131 | |
BC143347.1 | - | 24 - 25 | 2926 | 131 | |
DQ139833.1 | - | 23 - 24 | 2878 | 131 |