Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF10L
Entrez Gene ID
55160
Gene Name
Rho guanine nucleotide exchange factor 10 like
Gene Aliases
GrinchGEF
Chromosome Location
Chr.1: 17520197 - 17697875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011722.2 NP_001011722.2 1 - 2 78 73
NM_001319837.1 NP_001306766.1 1 - 2 78 73
NM_018125.3 NP_060595.3 2 - 3 194 73
NR_137287.1 2 - 3 194 73
NR_137288.1 2 - 3 194 73
XM_005245923.2 XP_005245980.2 2 - 3 158 73
XM_005245925.2 XP_005245982.2 2 - 3 158 73
XM_006710728.1 XP_006710791.1 2 - 3 340 73
XM_006710729.1 XP_006710792.1 2 - 3 158 73
XM_006710731.1 XP_006710794.1 2 - 3 340 73
XM_011541691.2 XP_011539993.1 1 - 2 164 73
XM_011541692.2 XP_011539994.1 1 - 2 164 73
XM_011541693.2 XP_011539995.1 1 - 2 164 73
XM_011541695.2 XP_011539997.1 1 - 2 164 73
XM_011541696.2 XP_011539998.1 1 - 2 164 73
XM_017001617.1 XP_016857106.1 2 - 3 158 73
XM_017001619.1 XP_016857108.1 1 - 2 164 73
XM_017001620.1 XP_016857109.1 1 - 2 164 73
XM_017001621.1 XP_016857110.1 1 - 2 177 73
XM_017001622.1 XP_016857111.1 1 - 2 177 73
GenBank mRNA AB046846.1 - 2 - 3 194 73
AK311076.1 - 2 - 3 121 73
BC117171.1 - 1 - 2 68 73
BC143347.1 - 1 - 2 68 73

View Full Product Details