Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIPA1L1
Entrez Gene ID
26037
Gene Name
signal induced proliferation associated 1 like 1
Gene Aliases
E6TP1
Chromosome Location
Chr.14: 71320418 - 71741229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284245.1 NP_001271174.1 14 - 15 4575 67
NM_001284246.1 NP_001271175.1 14 - 15 5164 67
NM_001284247.1 NP_001271176.1 14 - 15 4579 67
NM_015556.2 NP_056371.1 15 - 16 4638 67
XM_005267516.4 XP_005267573.1 15 - 16 5227 67
XM_006720111.2 XP_006720174.1 17 - 18 6517 67
XM_011536630.1 XP_011534932.1 19 - 20 4989 67
XM_011536631.1 XP_011534933.1 19 - 20 5400 67
XM_011536632.1 XP_011534934.1 18 - 19 4909 67
XM_011536635.2 XP_011534937.1 18 - 19 5056 67
XM_011536638.2 XP_011534940.1 14 - 15 3712 67
XM_017021159.1 XP_016876648.1 21 - 22 5146 67
XM_017021160.1 XP_016876649.1 19 - 20 5068 67
XM_017021161.1 XP_016876650.1 20 - 21 5021 67
XM_017021162.1 XP_016876651.1 22 - 23 5203 67
XM_017021163.1 XP_016876652.1 17 - 18 4870 67
XM_017021164.1 XP_016876653.1 17 - 18 4877 67
XM_017021165.1 XP_016876654.1 19 - 20 4946 67
XM_017021166.1 XP_016876655.1 20 - 21 5079 67
XM_017021167.1 XP_016876656.1 19 - 20 4954 67
XM_017021168.1 XP_016876657.1 19 - 20 4928 67
XM_017021169.1 XP_016876658.1 18 - 19 5081 67
XM_017021170.1 XP_016876659.1 18 - 19 5439 67
XM_017021171.1 XP_016876660.1 19 - 20 5012 67
XM_017021172.1 XP_016876661.1 18 - 19 4887 67
XM_017021173.1 XP_016876662.1 17 - 18 5014 67
XM_017021174.1 XP_016876663.1 17 - 18 5254 67
XM_017021175.1 XP_016876664.1 17 - 18 5372 67
XM_017021176.1 XP_016876665.1 22 - 23 5199 67
XM_017021177.1 XP_016876666.1 22 - 23 5188 67
XM_017021178.1 XP_016876667.1 20 - 21 5026 67
XM_017021179.1 XP_016876668.1 19 - 20 4929 67
XM_017021180.1 XP_016876669.1 23 - 24 5241 67
XM_017021181.1 XP_016876670.1 21 - 22 5146 67
XM_017021182.1 XP_016876671.1 20 - 21 5079 67
XM_017021183.1 XP_016876672.1 18 - 19 4887 67
XM_017021184.1 XP_016876673.1 22 - 23 5199 67
XM_017021185.1 XP_016876674.1 15 - 16 5227 67
XM_017021186.1 XP_016876675.1 20 - 21 5083 67
XM_017021187.1 XP_016876676.1 18 - 19 4949 67
XM_017021188.1 XP_016876677.1 17 - 18 4824 67
XM_017021189.1 XP_016876678.1 19 - 20 4933 67
XM_017021190.1 XP_016876679.1 20 - 21 5083 67
XM_017021191.1 XP_016876680.1 18 - 19 4891 67
XM_017021192.1 XP_016876681.1 17 - 18 4824 67
XM_017021193.1 XP_016876682.1 17 - 18 4822 67
XM_017021194.1 XP_016876683.1 20 - 21 4999 67
XM_017021195.1 XP_016876684.1 19 - 20 4936 67
XM_017021196.1 XP_016876685.1 19 - 20 4936 67
XM_017021197.1 XP_016876686.1 20 - 21 3346 67
XM_017021198.1 XP_016876687.1 17 - 18 3087 67
XM_017021199.1 XP_016876688.1 21 - 22 3388 67
XM_017021200.1 XP_016876689.1 17 - 18 3091 67
GenBank mRNA AB007900.2 - 14 - 15 4362 67
AF090989.1 - 14 - 15 4562 67
AF090990.1 - 15 - 16 4625 67
AK122930.1 - 14 - 15 5164 67
AK302650.1 - 13 - 14 2814 67
BC146777.1 - 14 - 15 4362 67

View Full Product Details