Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258008.1 NP_001244937.1 3 - 4 411 112
NM_017984.4 NP_060454.3 3 - 4 411 112
XM_005250479.2 XP_005250536.1 3 - 4 842 112
XM_005250480.2 XP_005250537.1 3 - 4 842 112
XM_006716035.3 XP_006716098.1 3 - 4 842 112
XM_006716036.2 XP_006716099.1 3 - 4 346 112
XM_006716037.3 XP_006716100.1 3 - 4 842 112
XM_006716038.3 XP_006716101.1 3 - 4 842 112
XM_006716040.3 XP_006716103.1 3 - 4 842 112
XM_017012379.1 XP_016867868.1 3 - 4 842 112
XM_017012380.1 XP_016867869.1 3 - 4 346 112
GenBank mRNA AK093038.1 - 3 - 4 346 112
AK299989.1 - 3 - 4 299 112
AK300745.1 - 3 - 4 267 112
AK302108.1 - 3 - 4 265 112
AK316553.1 - 3 - 4 599 112
AL136735.1 - 3 - 4 282 112

View Full Product Details