Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258008.1 NP_001244937.1 6 - 7 854 93
NM_017984.4 NP_060454.3 6 - 7 854 93
XM_005250479.2 XP_005250536.1 6 - 7 1285 93
XM_005250480.2 XP_005250537.1 6 - 7 1285 93
XM_006716035.3 XP_006716098.1 6 - 7 1285 93
XM_006716036.2 XP_006716099.1 6 - 7 789 93
XM_006716037.3 XP_006716100.1 6 - 7 1285 93
XM_006716038.3 XP_006716101.1 6 - 7 1285 93
XM_006716040.3 XP_006716103.1 6 - 7 1285 93
XM_017012379.1 XP_016867868.1 6 - 7 1285 93
XM_017012380.1 XP_016867869.1 6 - 7 789 93
GenBank mRNA AK000919.1 - 1 - 2 238 93
AK093038.1 - 6 - 7 789 93
AK300745.1 - 6 - 7 710 93
AK302108.1 - 5 - 6 588 93
AL136735.1 - 6 - 7 725 93
BC002725.1 - 3 - 4 552 93
KU178677.1 - 1 - 2 114 93
KU178678.1 - 1 - 2 114 93
KU178679.1 - 1 - 2 114 93
KU178680.1 - 1 - 2 114 93
KU178681.1 - 1 - 2 114 93
KU178682.1 - 1 - 2 114 93

View Full Product Details