Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr.7: 100400826 - 100429239 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017984.4 NP_060454.3 7 - 8 1013 100
XM_006716038.3 XP_006716101.1 7 - 8 1444 100
GenBank mRNA AL136735.1 - 7 - 8 884 100

View Full Product Details