Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC38A6
Entrez Gene ID
145389
Gene Name
solute carrier family 38 member 6
Gene Aliases
NAT-1, SNAT6
Chromosome Location
Chr.14: 60981114 - 61083733 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153811.2 NP_722518.2 15 - 16 1450 138
XM_006720050.1 XP_006720113.1 11 - 12 893 138
GenBank mRNA AF070578.1 - 17 - 18 1588 138
AF543422.1 - 15 - 16 1402 138
BC110378.1 - 15 - 16 1311 138
BX648332.1 - 15 - 16 1324 138

View Full Product Details