Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIOB
Entrez Gene ID
254528
Gene Name
meiosis specific with OB domains
Gene Aliases
C16orf73, gs129
Chromosome Location
Chr.16: 1833983 - 1872178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163560.2 NP_001157032.1 6 - 7 725 80
NM_152764.2 NP_689977.2 6 - 7 648 80
GenBank mRNA BC029829.1 - 2 - 3 250 80
CD300613.1 - 3 - 4 403 80

View Full Product Details