The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181698.3 | NP_859049.2 | 1 - 2 | 152 | 104 |
XM_011519358.1 | XP_011517660.1 | 1 - 2 | 112 | 104 | |
XM_011519360.2 | XP_011517662.1 | 1 - 2 | 106 | 104 | |
XM_011519361.1 | XP_011517663.1 | 1 - 2 | 109 | 104 | |
XM_011519362.1 | XP_011517664.1 | 1 - 2 | 152 | 104 | |
XM_011519363.1 | XP_011517665.1 | 1 - 2 | 97 | 104 | |
XM_011519364.1 | XP_011517666.1 | 1 - 2 | 108 | 104 | |
XM_011519365.2 | XP_011517667.1 | 1 - 2 | 106 | 104 | |
XM_011519366.1 | XP_011517668.1 | 1 - 2 | 109 | 104 | |
GenBank mRNA | AF429969.1 | - | 1 - 2 | 108 | 104 |
AK057280.1 | - | 1 - 2 | 108 | 104 | |
DB037034.1 | - | 1 - 2 | 152 | 104 |