The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003794.3 | NP_003785.1 | 3 - 4 | 445 | 63 |
NR_073435.1 | 2 - 3 | 323 | 63 | ||
XM_017007414.1 | XP_016862903.1 | 3 - 4 | 437 | 63 | |
GenBank mRNA | AF065485.1 | - | 3 - 4 | 403 | 63 |
AK001835.1 | - | 3 - 4 | 407 | 63 | |
AK298972.1 | - | 2 - 3 | 290 | 63 | |
BC018762.1 | - | 3 - 4 | 421 | 63 | |
BI669801.1 | - | 3 - 4 | 449 | 63 | |
DA289870.1 | - | 3 - 4 | 447 | 63 |