Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX4
Entrez Gene ID
8723
Gene Name
sorting nexin 4
Gene Aliases
ATG24B
Chromosome Location
Chr.3: 125446644 - 125520214 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003794.3 NP_003785.1 6 - 7 696 108
NR_073435.1 5 - 6 574 108
XM_017007414.1 XP_016862903.1 6 - 7 688 108
GenBank mRNA AF130078.1 - 1 - 2 63 108
AK001835.1 - 6 - 7 658 108
AK298972.1 - 5 - 6 541 108
BC018762.1 - 6 - 7 672 108

View Full Product Details