Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEZ6L
Entrez Gene ID
23544
Gene Name
seizure related 6 homolog like
Gene Aliases
-
Chromosome Location
Chr.22: 26169474 - 26383597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184773.1 NP_001171702.1 11 - 12 2596 130
NM_001184774.1 NP_001171703.1 11 - 12 2596 130
NM_001184775.1 NP_001171704.1 11 - 12 2596 130
NM_001184777.1 NP_001171706.1 11 - 12 2596 130
NM_021115.4 NP_066938.2 11 - 12 2596 130
XM_005261439.3 XP_005261496.1 11 - 12 2572 130
XM_006724195.3 XP_006724258.1 11 - 12 2572 130
XM_011530037.2 XP_011528339.1 11 - 12 2572 130
XM_011530038.2 XP_011528340.1 11 - 12 2572 130
XM_011530039.2 XP_011528341.1 11 - 12 2572 130
GenBank mRNA AB041736.1 - 11 - 12 2400 130
AK294790.1 - 1 - 2 192 130
AL035545.1 - 10 - 11 2461 130
AL050253.1 - 10 - 11 2461 130
AY358405.1 - 11 - 12 2596 130
BC126115.1 - 11 - 12 2495 130
BC143851.1 - 11 - 12 2495 130
BC143854.1 - 11 - 12 2495 130
CR456574.1 - 11 - 12 2412 130

View Full Product Details