Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEZ6L
Entrez Gene ID
23544
Gene Name
seizure related 6 homolog like
Gene Aliases
-
Chromosome Location
Chr.22: 26169474 - 26383597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184773.1 NP_001171702.1 16 - 17 3236 85
NM_001184774.1 NP_001171703.1 15 - 16 3206 85
NM_001184775.1 NP_001171704.1 15 - 16 3200 85
NM_001184776.1 NP_001171705.1 14 - 15 3014 85
NM_001184777.1 NP_001171706.1 14 - 15 3011 85
NM_021115.4 NP_066938.2 16 - 17 3239 85
XM_005261439.3 XP_005261496.1 15 - 16 3179 85
XM_005261440.3 XP_005261497.1 15 - 16 3024 85
XM_006724195.3 XP_006724258.1 16 - 17 3209 85
XM_011530037.2 XP_011528339.1 15 - 16 3020 85
XM_011530038.2 XP_011528340.1 15 - 16 3017 85
XM_011530039.2 XP_011528341.1 15 - 16 3014 85
XM_017028699.1 XP_016884188.1 15 - 16 3021 85
XM_017028700.1 XP_016884189.1 15 - 16 3018 85
GenBank mRNA AB023144.2 - 14 - 15 2974 85
AK294790.1 - 4 - 5 687 85
AY358405.1 - 16 - 17 3236 85
BC126115.1 - 16 - 17 3138 85
BC143851.1 - 15 - 16 3105 85
BC143854.1 - 15 - 16 3099 85
BM875686.1 - 6 - 7 518 85
CR456574.1 - 14 - 15 2827 85

View Full Product Details