Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEZ6L
Entrez Gene ID
23544
Gene Name
seizure related 6 homolog like
Gene Aliases
-
Chromosome Location
Chr.22: 26169474 - 26383597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184773.1 NP_001171702.1 4 - 5 1361 84
NM_001184774.1 NP_001171703.1 4 - 5 1361 84
NM_001184775.1 NP_001171704.1 4 - 5 1361 84
NM_001184776.1 NP_001171705.1 4 - 5 1361 84
NM_001184777.1 NP_001171706.1 4 - 5 1361 84
NM_021115.4 NP_066938.2 4 - 5 1361 84
XM_005261439.3 XP_005261496.1 4 - 5 1337 84
XM_005261440.3 XP_005261497.1 4 - 5 1338 84
XM_006724195.3 XP_006724258.1 4 - 5 1337 84
XM_011530037.2 XP_011528339.1 4 - 5 1337 84
XM_011530038.2 XP_011528340.1 4 - 5 1337 84
XM_011530039.2 XP_011528341.1 4 - 5 1337 84
XM_017028699.1 XP_016884188.1 4 - 5 1338 84
XM_017028700.1 XP_016884189.1 4 - 5 1338 84
GenBank mRNA AB023144.2 - 4 - 5 1321 84
AB041736.1 - 4 - 5 1165 84
AL035545.1 - 3 - 4 1226 84
AL050253.1 - 3 - 4 1226 84
AY358405.1 - 4 - 5 1361 84
BC126115.1 - 4 - 5 1260 84
BC143851.1 - 4 - 5 1260 84
BC143854.1 - 4 - 5 1260 84
CR456574.1 - 4 - 5 1177 84

View Full Product Details