The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184773.1 | NP_001171702.1 | 10 - 11 | 2410 | 79 |
NM_001184774.1 | NP_001171703.1 | 10 - 11 | 2410 | 79 | |
NM_001184775.1 | NP_001171704.1 | 10 - 11 | 2410 | 79 | |
NM_001184776.1 | NP_001171705.1 | 10 - 11 | 2410 | 79 | |
NM_001184777.1 | NP_001171706.1 | 10 - 11 | 2410 | 79 | |
NM_021115.4 | NP_066938.2 | 10 - 11 | 2410 | 79 | |
XM_005261439.3 | XP_005261496.1 | 10 - 11 | 2386 | 79 | |
XM_005261440.3 | XP_005261497.1 | 10 - 11 | 2387 | 79 | |
XM_006724195.3 | XP_006724258.1 | 10 - 11 | 2386 | 79 | |
XM_011530037.2 | XP_011528339.1 | 10 - 11 | 2386 | 79 | |
XM_011530038.2 | XP_011528340.1 | 10 - 11 | 2386 | 79 | |
XM_011530039.2 | XP_011528341.1 | 10 - 11 | 2386 | 79 | |
XM_017028699.1 | XP_016884188.1 | 10 - 11 | 2387 | 79 | |
XM_017028700.1 | XP_016884189.1 | 10 - 11 | 2387 | 79 | |
GenBank mRNA | AB023144.2 | - | 10 - 11 | 2370 | 79 |
AB041736.1 | - | 10 - 11 | 2214 | 79 | |
AL035545.1 | - | 9 - 10 | 2275 | 79 | |
AL050253.1 | - | 9 - 10 | 2275 | 79 | |
AY358405.1 | - | 10 - 11 | 2410 | 79 | |
BC126115.1 | - | 10 - 11 | 2309 | 79 | |
BC143851.1 | - | 10 - 11 | 2309 | 79 | |
BC143854.1 | - | 10 - 11 | 2309 | 79 | |
CR456574.1 | - | 10 - 11 | 2226 | 79 |