Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G3
Entrez Gene ID
8672
Gene Name
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location
Chr.1: 20806292 - 21177320 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198801.1 NP_001185730.1 14 - 15 2587 65
NM_001198802.1 NP_001185731.1 15 - 16 2608 65
NM_003760.4 NP_003751.2 12 - 13 2224 65
XM_011542356.2 XP_011540658.1 17 - 18 3187 65
XM_011542361.2 XP_011540663.1 17 - 18 3187 65
XM_011542370.1 XP_011540672.1 17 - 18 3190 65
XM_011542371.1 XP_011540673.1 15 - 16 2932 65
XM_011542374.1 XP_011540676.1 14 - 15 2899 65
XM_011542375.2 XP_011540677.1 14 - 15 4487 65
XM_011542381.2 XP_011540683.1 4 - 5 573 65
XM_017002672.1 XP_016858161.1 18 - 19 4725 65
XM_017002673.1 XP_016858162.1 16 - 17 3169 65
XM_017002674.1 XP_016858163.1 16 - 17 3166 65
XM_017002675.1 XP_016858164.1 16 - 17 3157 65
XM_017002676.1 XP_016858165.1 15 - 16 3136 65
XM_017002677.1 XP_016858166.1 15 - 16 3013 65
XM_017002678.1 XP_016858167.1 17 - 18 3190 65
XM_017002679.1 XP_016858168.1 16 - 17 3076 65
XM_017002680.1 XP_016858169.1 13 - 14 2106 65
XM_017002681.1 XP_016858170.1 16 - 17 3073 65
XM_017002682.1 XP_016858171.1 16 - 17 3166 65
XM_017002683.1 XP_016858172.1 15 - 16 3055 65
XM_017002684.1 XP_016858173.1 16 - 17 4581 65
XM_017002685.1 XP_016858174.1 16 - 17 3157 65
XM_017002686.1 XP_016858175.1 16 - 17 3154 65
XM_017002687.1 XP_016858176.1 15 - 16 3043 65
XM_017002688.1 XP_016858177.1 12 - 13 2072 65
XM_017002689.1 XP_016858178.1 17 - 18 3190 65
XM_017002690.1 XP_016858179.1 15 - 16 3040 65
XM_017002691.1 XP_016858180.1 15 - 16 3136 65
XM_017002692.1 XP_016858181.1 14 - 15 3022 65
XM_017002693.1 XP_016858182.1 13 - 14 2893 65
XM_017002694.1 XP_016858183.1 16 - 17 3076 65
XM_017002695.1 XP_016858184.1 16 - 17 3073 65
XM_017002696.1 XP_016858185.1 15 - 16 3055 65
XM_017002697.1 XP_016858186.1 15 - 16 3052 65
XM_017002698.1 XP_016858187.1 15 - 16 3043 65
XM_017002699.1 XP_016858188.1 17 - 18 3190 65
XM_017002700.1 XP_016858189.1 14 - 15 3022 65
XM_017002701.1 XP_016858190.1 14 - 15 3019 65
XM_017002702.1 XP_016858191.1 13 - 14 2842 65
XM_017002703.1 XP_016858192.1 12 - 13 2114 65
XM_017002704.1 XP_016858193.1 16 - 17 3166 65
XM_017002705.1 XP_016858194.1 16 - 17 2350 65
XM_017002706.1 XP_016858195.1 16 - 17 2350 65
XM_017002707.1 XP_016858196.1 13 - 14 2182 65
XM_017002708.1 XP_016858197.1 13 - 14 2182 65
XM_017002709.1 XP_016858198.1 14 - 15 2934 65
XM_017002710.1 XP_016858199.1 17 - 18 3190 65
XM_017002711.1 XP_016858200.1 17 - 18 3190 65
GenBank mRNA AB209119.1 - 15 - 16 2567 65
AF012072.2 - 12 - 13 2224 65
AK294883.1 - 6 - 7 1471 65
AK302873.1 - 2 - 3 354 65
AK310195.1 - 2 - 3 196 65
BC094683.1 - 10 - 11 1231 65
BC136643.1 - 12 - 13 2147 65

View Full Product Details