Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EIF4G3
- Entrez Gene ID
-
8672
- Gene Name
- eukaryotic translation initiation factor 4 gamma 3
- Gene Aliases
-
eIF-4G 3, eIF4G 3, eIF4GII
- Chromosome Location
-
Chr.1: 20806292 - 21177320 on Build GRCh38
View Full Product Details