Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKAL1
Entrez Gene ID
54901
Gene Name
CDK5 regulatory subunit associated protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.6: 20534457 - 21232404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017774.3 NP_060244.2 14 - 15 1546 138
XM_017010987.1 XP_016866476.1 13 - 14 1439 138
GenBank mRNA AK000349.1 - 14 - 15 1484 138
AK291735.1 - 14 - 15 1526 138
BC121020.1 - 14 - 15 1476 138
BC121021.1 - 14 - 15 1476 138

View Full Product Details