The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002742.2 | NP_002733.2 | 8 - 9 | 1499 | 94 |
XM_005267859.1 | XP_005267916.1 | 9 - 10 | 1457 | 94 | |
XM_011536964.2 | XP_011535266.1 | 9 - 10 | 3879 | 94 | |
XM_011536965.2 | XP_011535267.1 | 9 - 10 | 5929 | 94 | |
XM_017021461.1 | XP_016876950.1 | 9 - 10 | 1234 | 94 | |
XM_017021462.1 | XP_016876951.1 | 9 - 10 | 1196 | 94 | |
GenBank mRNA | AK314170.1 | - | 8 - 9 | 1435 | 94 |
X75756.1 | - | 8 - 9 | 1553 | 94 |