Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NPAS3
- Entrez Gene ID
-
64067
- Gene Name
- neuronal PAS domain protein 3
- Gene Aliases
-
MOP6, PASD6, bHLHe12
- Chromosome Location
-
Chr.14: 32934785 - 33804176 on Build GRCh38
View Full Product Details