Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPAS3
Entrez Gene ID
64067
Gene Name
neuronal PAS domain protein 3
Gene Aliases
MOP6, PASD6, bHLHe12
Chromosome Location
Chr.14: 32934785 - 33804176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164749.1 NP_001158221.1 8 - 9 1101 129
NM_001165893.1 NP_001159365.1 7 - 8 1011 129
NM_022123.2 NP_071406.1 7 - 8 1005 129
NM_173159.2 NP_775182.1 8 - 9 1062 129
XM_005267991.3 XP_005268048.2 8 - 9 1206 129
XM_005267992.3 XP_005268049.2 8 - 9 1188 129
XM_011537067.2 XP_011535369.1 9 - 10 1202 129
XM_011537069.2 XP_011535371.2 8 - 9 1206 129
XM_011537070.2 XP_011535372.1 8 - 9 1106 129
XM_011537071.2 XP_011535373.2 7 - 8 1101 129
XM_011537072.2 XP_011535374.1 7 - 8 1060 129
XM_017021582.1 XP_016877071.1 9 - 10 1257 129
XM_017021583.1 XP_016877072.1 9 - 10 1257 129
XM_017021584.1 XP_016877073.1 8 - 9 1167 129
XM_017021585.1 XP_016877074.1 7 - 8 1116 129
XM_017021586.1 XP_016877075.1 7 - 8 4013 129
XM_017021587.1 XP_016877076.1 7 - 8 3255 129
XM_017021588.1 XP_016877077.1 8 - 9 1203 129
GenBank mRNA AB054575.1 - 7 - 8 992 129
AB054576.1 - 8 - 9 998 129
AB055962.1 - 8 - 9 1037 129
AF164438.1 - 7 - 8 1016 129
AY157302.1 - 7 - 8 1005 129
KJ534893.1 - 7 - 8 941 129
KJ534894.1 - 9 - 10 1037 129
KJ534895.1 - 7 - 8 947 129
KJ534896.1 - 7 - 8 941 129
KJ534897.1 - 8 - 9 992 129
KJ534898.1 - 7 - 8 947 129
KJ535019.1 - 8 - 9 998 129

View Full Product Details