The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184937.1 | NP_001171866.1 | 8 - 9 | 822 | 117 |
NM_001184938.2 | NP_001171867.1 | 8 - 9 | 1112 | 117 | |
NM_001184939.1 | NP_001171868.1 | 8 - 9 | 822 | 117 | |
NM_020909.3 | NP_065960.2 | 8 - 9 | 822 | 117 | |
NR_135920.1 | 8 - 9 | 822 | 117 | ||
XM_006712651.1 | XP_006712714.1 | 8 - 9 | 802 | 117 | |
XM_006712652.1 | XP_006712715.1 | 8 - 9 | 1117 | 117 | |
XM_006712653.1 | XP_006712716.1 | 8 - 9 | 822 | 117 | |
XM_011511529.2 | XP_011509831.1 | 8 - 9 | 802 | 117 | |
XM_017004567.1 | XP_016860056.1 | 8 - 9 | 658 | 117 | |
XM_017004568.1 | XP_016860057.1 | 8 - 9 | 822 | 117 | |
GenBank mRNA | AB046768.1 | - | 1 - 2 | 71 | 117 |
AK023019.1 | - | 8 - 9 | 773 | 117 | |
AK290895.1 | - | 8 - 9 | 732 | 117 | |
BC032822.2 | - | 8 - 9 | 1112 | 117 | |
BC054508.1 | - | 8 - 9 | 757 | 117 |