The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000122.1 | NP_000113.1 | 13 - 14 | 2156 | 67 |
NM_001303416.1 | NP_001290345.1 | 13 - 14 | 2271 | 67 | |
NM_001303418.1 | NP_001290347.1 | 13 - 14 | 2333 | 67 | |
XM_011510794.1 | XP_011509096.1 | 13 - 14 | 2135 | 67 | |
XM_011510795.1 | XP_011509097.1 | 13 - 14 | 2009 | 67 | |
XM_017003583.1 | XP_016859072.1 | 13 - 14 | 1991 | 67 | |
GenBank mRNA | AK091500.1 | - | 13 - 14 | 2258 | 67 |
AK095557.1 | - | 13 - 14 | 2333 | 67 | |
AK127469.1 | - | 9 - 10 | 4032 | 67 | |
AK290474.1 | - | 13 - 14 | 2082 | 67 | |
BC008820.2 | - | 13 - 14 | 2125 | 67 | |
M31899.1 | - | 13 - 14 | 2156 | 67 |