The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001223.4 | NP_001214.1 | 5 - 6 | 842 | 126 |
NM_001257118.2 | NP_001244047.1 | 6 - 7 | 905 | 126 | |
NM_001257119.2 | NP_001244048.1 | 5 - 6 | 842 | 126 | |
NM_033292.3 | NP_150634.1 | 6 - 7 | 905 | 126 | |
NM_033293.3 | NP_150635.1 | 5 - 6 | 626 | 126 | |
XM_017018393.1 | XP_016873882.1 | 6 - 7 | 1253 | 126 | |
XM_017018394.1 | XP_016873883.1 | 6 - 7 | 1253 | 126 | |
XM_017018395.1 | XP_016873884.1 | 5 - 6 | 1190 | 126 | |
XM_017018396.1 | XP_016873885.1 | 6 - 7 | 1162 | 126 | |
GenBank mRNA | AK223503.1 | - | 6 - 7 | 897 | 126 |
AK290122.1 | - | 5 - 6 | 833 | 126 | |
AK301037.1 | - | 6 - 7 | 805 | 126 | |
AK310059.1 | - | 4 - 5 | 716 | 126 | |
AK313516.1 | - | 6 - 7 | 891 | 126 | |
AY660536.1 | - | 6 - 7 | 799 | 126 | |
BC041689.2 | - | 4 - 5 | 791 | 126 | |
BC062327.1 | - | 5 - 6 | 592 | 126 | |
M87507.1 | - | 6 - 7 | 878 | 126 | |
U13697.1 | - | 4 - 5 | 798 | 126 | |
U13698.1 | - | 4 - 5 | 582 | 126 | |
X65019.1 | - | 5 - 6 | 868 | 126 |