The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282415.1 | NP_001269344.1 | 2 - 3 | 403 | 59 |
NM_032805.2 | NP_116194.2 | 3 - 4 | 866 | 59 | |
XM_017023791.1 | XP_016879280.1 | 2 - 3 | 902 | 59 | |
XM_017023792.1 | XP_016879281.1 | 2 - 3 | 902 | 59 | |
GenBank mRNA | AK027455.1 | - | 2 - 3 | 594 | 59 |
AK074736.1 | - | 2 - 3 | 378 | 59 | |
BC111367.1 | - | 1 - 2 | 1195 | 59 |