Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
Chromosome Location
Chr.10: 59245933 - 59363186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001971.2 NP_001001971.1 2 - 3 256 94
NM_001143773.1 NP_001137245.1 3 - 4 427 94
NM_001166698.1 NP_001160170.1 3 - 4 341 94
NM_198215.3 NP_937858.2 2 - 3 256 94
XM_005269618.4 XP_005269675.2 2 - 3 256 94
XM_005269619.4 XP_005269676.2 2 - 3 256 94
XM_006717702.3 XP_006717765.1 4 - 5 257 94
XM_006717703.3 XP_006717766.1 4 - 5 439 94
XM_011539490.2 XP_011537792.1 2 - 3 223 94
XM_011539491.2 XP_011537793.1 3 - 4 269 94
XM_017015885.1 XP_016871374.1 3 - 4 170 94
XM_017015886.1 XP_016871375.1 3 - 4 427 94
XM_017015887.1 XP_016871376.1 3 - 4 177 94
XM_017015888.1 XP_016871377.1 4 - 5 439 94
XM_017015889.1 XP_016871378.1 4 - 5 257 94
XM_017015890.1 XP_016871379.1 3 - 4 407 94
XM_017015891.1 XP_016871380.1 3 - 4 427 94
XM_017015892.1 XP_016871381.1 3 - 4 427 94
XM_017015893.1 XP_016871382.1 4 - 5 439 94
GenBank mRNA AK294805.1 - 2 - 3 171 94
AK316542.1 - 3 - 4 337 94
BC036453.1 - 2 - 3 214 94
BC064431.1 - 2 - 3 256 94
DA188937.1 - 2 - 3 256 94

View Full Product Details