Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM5
Entrez Gene ID
10181
Gene Name
RNA binding motif protein 5
Gene Aliases
G15, H37, LUCA15, RMB5
Chromosome Location
Chr.3: 50088908 - 50118964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005778.3 NP_005769.1 11 - 12 1125 102
NR_036627.2 10 - 11 1051 102
XM_006712917.1 XP_006712980.1 11 - 12 1125 102
XM_011533261.1 XP_011531563.1 9 - 10 903 102
XM_011533262.1 XP_011531564.1 11 - 12 1125 102
XM_017005503.1 XP_016860992.1 11 - 12 1107 102
XM_017005504.1 XP_016860993.1 9 - 10 903 102
XM_017005505.1 XP_016860994.1 10 - 11 1018 102
XM_017005506.1 XP_016860995.1 8 - 9 868 102
XM_017005507.1 XP_016860996.1 7 - 8 3899 102
GenBank mRNA AF091263.1 - 11 - 12 1098 102
AF103802.4 - 11 - 12 1076 102
AK297249.1 - 10 - 11 1005 102
AK297445.1 - 11 - 12 980 102
AK302766.1 - 10 - 11 933 102
AK314032.1 - 11 - 12 1080 102
AU129811.1 - 4 - 5 392 102
BC002957.1 - 11 - 12 1071 102
DA776815.1 - 4 - 5 346 102
KU178505.1 - 10 - 11 950 102
KU178506.1 - 9 - 10 876 102
U23946.1 - 11 - 12 1019 102

View Full Product Details