Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP3
Entrez Gene ID
55240
Gene Name
STEAP3 metalloreductase
Gene Aliases
AHMIO2, STMP3, TSAP6, dudlin-2, dudulin-2, pHyde
Chromosome Location
Chr.2: 119222476 - 119265652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008410.1 NP_001008410.1 2 - 3 582 66
NM_018234.2 NP_060704.2 3 - 4 650 66
NM_138637.2 NP_619543.2 2 - 3 582 66
NM_182915.2 NP_878919.2 3 - 4 997 66
XM_006712614.3 XP_006712677.1 3 - 4 724 66
XM_006712615.1 XP_006712678.1 2 - 3 594 66
XM_011511403.1 XP_011509705.1 4 - 5 1008 66
GenBank mRNA AF423424.1 - 3 - 4 627 66
AK001691.1 - 3 - 4 627 66
AK291608.1 - 3 - 4 959 66
AL833624.1 - 3 - 4 997 66
AY029585.1 - 3 - 4 627 66
AY082673.1 - 2 - 3 537 66
AY214461.1 - 3 - 4 627 66
BC042150.1 - 2 - 3 533 66
BC095421.1 - 2 - 3 537 66
BX538047.1 - 3 - 4 966 66

View Full Product Details