The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136154.1 | NP_001129626.1 | 8 - 9 | 1040 | 111 |
NM_001136155.1 | NP_001129627.1 | 5 - 6 | 653 | 111 | |
NM_001243428.1 | NP_001230357.1 | 8 - 9 | 1063 | 111 | |
NM_001243432.2 | NP_001230361.1 | 8 - 9 | 1040 | 111 | |
NM_182918.3 | NP_891548.1 | 6 - 7 | 871 | 111 | |
XM_017028287.1 | XP_016883776.1 | 7 - 8 | 1224 | 111 | |
GenBank mRNA | AK297807.1 | - | 8 - 9 | 1040 | 111 |
AK300395.1 | - | 6 - 7 | 861 | 111 | |
AK301277.1 | - | 4 - 5 | 397 | 111 | |
AK303518.1 | - | 5 - 6 | 626 | 111 | |
AK309469.1 | - | 8 - 9 | 1063 | 111 | |
AY204741.1 | - | 8 - 9 | 954 | 111 | |
BC040168.1 | - | 6 - 7 | 850 | 111 |