The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317381.1 | NP_001304310.1 | 7 - 8 | 992 | 135 |
NM_001317382.1 | NP_001304311.1 | 6 - 7 | 711 | 135 | |
NM_001317383.1 | NP_001304312.1 | 5 - 6 | 879 | 135 | |
NM_014278.3 | NP_055093.2 | 6 - 7 | 1002 | 135 | |
XM_011531745.2 | XP_011530047.1 | 6 - 7 | 1071 | 135 | |
GenBank mRNA | AB023421.1 | - | 6 - 7 | 714 | 135 |
AK027822.1 | - | 6 - 7 | 889 | 135 | |
AK302115.1 | - | 6 - 7 | 685 | 135 | |
AK303053.1 | - | 7 - 8 | 992 | 135 | |
AK307464.1 | - | 5 - 6 | 766 | 135 | |
AY290862.1 | - | 6 - 7 | 868 | 135 | |
BC040560.1 | - | 6 - 7 | 913 | 135 | |
EF197155.1 | - | 6 - 7 | 660 | 135 |