The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002395.5 | NP_002386.1 | 12 - 13 | 1714 | 88 |
XM_011535836.2 | XP_011534138.1 | 11 - 12 | 1439 | 88 | |
GenBank mRNA | AK223417.1 | - | 12 - 13 | 1540 | 88 |
AK289783.1 | - | 12 - 13 | 1542 | 88 | |
AK301875.1 | - | 10 - 11 | 1256 | 88 | |
AK302777.1 | - | 11 - 12 | 1439 | 88 | |
BC017403.1 | - | 3 - 4 | 434 | 88 | |
BC025246.1 | - | 12 - 13 | 1530 | 88 | |
U43944.1 | - | 12 - 13 | 1428 | 88 | |
X77244.1 | - | 12 - 13 | 1449 | 88 |