The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002395.5 | NP_002386.1 | 2 - 3 | 479 | 137 |
GenBank mRNA | AK223417.1 | - | 2 - 3 | 305 | 137 |
AK289783.1 | - | 2 - 3 | 307 | 137 | |
BC025246.1 | - | 2 - 3 | 295 | 137 | |
DA520723.1 | - | 2 - 3 | 479 | 137 | |
L34035.1 | - | 2 - 3 | 226 | 137 | |
U43944.1 | - | 2 - 3 | 193 | 137 | |
X77244.1 | - | 2 - 3 | 214 | 137 |