The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002395.5 | NP_002386.1 | 7 - 8 | 1078 | 83 |
XM_011535836.2 | XP_011534138.1 | 6 - 7 | 803 | 83 | |
GenBank mRNA | AK223417.1 | - | 7 - 8 | 904 | 83 |
AK289783.1 | - | 7 - 8 | 906 | 83 | |
AK301875.1 | - | 5 - 6 | 620 | 83 | |
AK302777.1 | - | 6 - 7 | 803 | 83 | |
BC025246.1 | - | 7 - 8 | 894 | 83 | |
L34035.1 | - | 7 - 8 | 825 | 83 | |
U43944.1 | - | 7 - 8 | 792 | 83 | |
X77244.1 | - | 7 - 8 | 813 | 83 |