Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK11
Entrez Gene ID
79858
Gene Name
NIMA related kinase 11
Gene Aliases
-
Chromosome Location
Chr.3: 131026850 - 131350465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146003.1 NP_001139475.1 10 - 11 1331 90
NM_001321220.1 NP_001308149.1 10 - 11 1331 90
NM_001321221.1 NP_001308150.1 12 - 13 1530 90
NM_001321222.1 NP_001308151.1 7 - 8 1016 90
NM_001321223.1 NP_001308152.1 10 - 11 1449 90
NM_001321224.1 NP_001308153.1 9 - 10 1165 90
NM_024800.4 NP_079076.3 11 - 12 1404 90
NM_145910.3 NP_665917.1 11 - 12 1404 90
XM_011513167.2 XP_011511469.1 11 - 12 1604 90
XM_011513173.1 XP_011511475.1 9 - 10 1080 90
XM_011513174.1 XP_011511476.1 11 - 12 1350 90
XM_011513175.1 XP_011511477.1 10 - 11 1192 90
XM_011513181.1 XP_011511483.1 7 - 8 733 90
XM_017007205.1 XP_016862694.1 12 - 13 1707 90
XM_017007206.1 XP_016862695.1 12 - 13 1625 90
XM_017007207.1 XP_016862696.1 12 - 13 1841 90
XM_017007208.1 XP_016862697.1 11 - 12 1425 90
XM_017007209.1 XP_016862698.1 11 - 12 1634 90
XM_017007210.1 XP_016862699.1 11 - 12 1490 90
XM_017007211.1 XP_016862700.1 11 - 12 1425 90
XM_017007212.1 XP_016862701.1 11 - 12 1403 90
XM_017007213.1 XP_016862702.1 11 - 12 1403 90
XM_017007214.1 XP_016862703.1 10 - 11 1506 90
XM_017007215.1 XP_016862704.1 11 - 12 1403 90
XM_017007216.1 XP_016862705.1 10 - 11 1299 90
XM_017007217.1 XP_016862706.1 8 - 9 1088 90
XM_017007218.1 XP_016862707.1 13 - 14 1608 90
XM_017007219.1 XP_016862708.1 10 - 11 1102 90
XM_017007220.1 XP_016862709.1 11 - 12 1425 90
XM_017007221.1 XP_016862710.1 11 - 12 1425 90
XM_017007222.1 XP_016862711.1 11 - 12 1425 90
XM_017007223.1 XP_016862712.1 7 - 8 984 90
XM_017007224.1 XP_016862713.1 12 - 13 1469 90
XM_017007225.1 XP_016862714.1 12 - 13 1482 90
XM_017007226.1 XP_016862715.1 12 - 13 1746 90
XM_017007227.1 XP_016862716.1 10 - 11 1226 90
XM_017007228.1 XP_016862717.1 8 - 9 841 90
XM_017007229.1 XP_016862718.1 8 - 9 1088 90
XM_017007230.1 XP_016862719.1 8 - 9 1266 90
XM_017007231.1 XP_016862720.1 10 - 11 1204 90
XM_017007232.1 XP_016862721.1 11 - 12 1409 90
XM_017007233.1 XP_016862722.1 10 - 11 1159 90
GenBank mRNA AB071996.1 - 11 - 12 1371 90
AB071997.1 - 11 - 12 1371 90
AK027148.1 - 6 - 7 597 90
AK293261.1 - 9 - 10 1080 90
AK297300.1 - 7 - 8 1016 90
AK316352.1 - 7 - 8 931 90
AL833472.1 - 10 - 11 1302 90
BC028587.2 - 10 - 11 1448 90
BX328072.2 - 4 - 5 399 90

View Full Product Details