Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK11
Entrez Gene ID
79858
Gene Name
NIMA related kinase 11
Gene Aliases
-
Chromosome Location
Chr.3: 131026850 - 131350465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146003.1 NP_001139475.1 14 - 15 1815 77
NM_001321220.1 NP_001308149.1 14 - 15 1815 77
NM_001321221.1 NP_001308150.1 16 - 17 2014 77
NM_001321222.1 NP_001308151.1 11 - 12 1500 77
NM_001321224.1 NP_001308153.1 12 - 13 1541 77
NM_024800.4 NP_079076.3 15 - 16 1888 77
XM_011513167.2 XP_011511469.1 15 - 16 2088 77
XM_011513173.1 XP_011511475.1 13 - 14 1564 77
XM_011513174.1 XP_011511476.1 15 - 16 1834 77
XM_011513175.1 XP_011511477.1 14 - 15 1676 77
XM_011513181.1 XP_011511483.1 11 - 12 1217 77
XM_017007205.1 XP_016862694.1 16 - 17 2191 77
XM_017007206.1 XP_016862695.1 16 - 17 2109 77
XM_017007207.1 XP_016862696.1 16 - 17 2325 77
XM_017007208.1 XP_016862697.1 15 - 16 1909 77
XM_017007209.1 XP_016862698.1 15 - 16 2118 77
XM_017007210.1 XP_016862699.1 15 - 16 1974 77
XM_017007211.1 XP_016862700.1 15 - 16 1909 77
XM_017007212.1 XP_016862701.1 14 - 15 1779 77
XM_017007213.1 XP_016862702.1 15 - 16 1887 77
XM_017007214.1 XP_016862703.1 14 - 15 1990 77
XM_017007215.1 XP_016862704.1 14 - 15 1779 77
XM_017007216.1 XP_016862705.1 13 - 14 1675 77
XM_017007217.1 XP_016862706.1 12 - 13 1572 77
XM_017007218.1 XP_016862707.1 17 - 18 2092 77
XM_017007219.1 XP_016862708.1 14 - 15 1586 77
XM_017007223.1 XP_016862712.1 10 - 11 1360 77
XM_017007224.1 XP_016862713.1 16 - 17 1953 77
XM_017007225.1 XP_016862714.1 16 - 17 1966 77
XM_017007226.1 XP_016862715.1 16 - 17 2230 77
XM_017007227.1 XP_016862716.1 14 - 15 1710 77
XM_017007228.1 XP_016862717.1 12 - 13 1325 77
XM_017007229.1 XP_016862718.1 12 - 13 1572 77
XM_017007230.1 XP_016862719.1 12 - 13 1750 77
XM_017007231.1 XP_016862720.1 13 - 14 1580 77
XM_017007232.1 XP_016862721.1 15 - 16 1893 77
XM_017007233.1 XP_016862722.1 14 - 15 1643 77
GenBank mRNA AB071996.1 - 15 - 16 1855 77
AK027148.1 - 10 - 11 1081 77
AK293261.1 - 12 - 13 1456 77
AK297300.1 - 11 - 12 1500 77
AK316352.1 - 11 - 12 1415 77
AL833472.1 - 14 - 15 1786 77

View Full Product Details