The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006454.2 | NP_006445.1 | 5 - 6 | 511 | 90 |
XM_011513388.2 | XP_011511690.1 | 6 - 7 | 1016 | 90 | |
GenBank mRNA | AF040963.1 | - | 5 - 6 | 492 | 90 |
AF082740.1 | - | 5 - 6 | 511 | 90 | |
AF187724.1 | - | 4 - 5 | 348 | 90 | |
AK024501.1 | - | 2 - 3 | 2997 | 90 | |
BC068060.1 | - | 5 - 6 | 564 | 90 | |
BT019969.1 | - | 5 - 6 | 479 | 90 | |
BT019970.1 | - | 5 - 6 | 479 | 90 |